Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NETO2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP162427
Description
NETO2 Polyclonal specifically detects NETO2 in Human samples. It is validated for Western Blot.Specifications
NETO2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 2, BTCL2, FLJ10430, FLJ90456, NEOT2FLJ14724, neuropilin (NRP) and tolloid (TLL)-like 2, neuropilin and tolloid-like protein 2 | |
Rabbit | |
57 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8NC67 | |
NETO2 | |
Synthetic peptides corresponding to NETO2(neuropilin (NRP) and tolloid (TLL)-like 2) The peptide sequence was selected from the N terminal of NETO2 (NP_060562). GIKHIPATQCGIWVRTSNGGHFASPNYPDSYPPNKECIYILEAAPRQRIE. | |
Affinity purified | |
RUO | |
81831 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction