Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NETO2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen NETO2
Immunogen Synthetic peptides corresponding to NETO2(neuropilin (NRP) and tolloid (TLL)-like 2) The peptide sequence was selected from the N terminal of NETO2. Peptide sequence ELSGADGIVRSSQVEQEEKTKPGQAVDCIWTIKATPKAKIYLRFLDYQME.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1625920 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16259120UL
20ul Each for $149.00
Add to cart
 
NBP162591 View Documents Novus BiologicalsSupplier Diversity Partner
NBP162591
100 ul Each for $329.00
Add to cart
 
Description

Description

NETO2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blotting.
Specifications

Specifications

NETO2
Rabbit
Polyclonal
Unconjugated
81831
Affinity Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
RUO
Synthetic peptides corresponding to NETO2(neuropilin (NRP) and tolloid (TLL)-like 2) The peptide sequence was selected from the N terminal of NETO2. Peptide sequence ELSGADGIVRSSQVEQEEKTKPGQAVDCIWTIKATPKAKIYLRFLDYQME.
Primary
IgG
Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 2, BTCL2, FLJ10430, FLJ90456, NEOT2FLJ14724, neuropilin (NRP) and tolloid (TLL)-like 2, neuropilin and tolloid-like protein 2
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit