Learn More
Netrin-2 Goat anti-Chicken, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF127SP
Catalog No. AF127SP
Add to cart
This item is not returnable. View return policy
Description
Description
Netrin-2 Polyclonal specifically detects Netrin-2 in Chicken samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
Specifications
Netrin-2
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Q90923
NTN3
Mouse myeloma cell line NS0-derived recombinant chicken Netrin-2 Ala16-Pro581 Accession # Q90923
Immunogen affinity purified
RUO
Primary
4917
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects chicken Netrin-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant chicken Netrin-1 is observed and less than 1% cross-reactivity with recombinant human Netrin-4, recombinant mouse (rm) Netrin-G1a and rmNetrin-G2a is observed.
Chicken