Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEU2 Mouse anti-Human, Clone: OTI8C12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503332

Catalog No. 50-167-4254


Add to cart

Description

Description

This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
Specifications

Specifications

NEU2
Monoclonal
0.93 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y3R4
SIAL2
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4759
Western Blot
OTI8C12
Unconjugated
NEU2
Liquid
NEU2
Full length human recombit protein of human NEU2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit