Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEU2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$104.00 - $345.50

Specifications

Antigen NEU2
Dilution Immunohistochemistry
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB120134
View Documents
Novus Biologicals
NBP31099325UL
25 μg
Each for $104.00
Add to cart
 
NB120133
View Documents
Novus Biologicals
NBP310993100UL
100 μg
Each for $345.50
Add to cart
 
Description

Description

NEU2 Polyclonal antibody specifically detects NEU2 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

NEU2
Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
4759
Polyclonal
Affinity purified
Immunohistochemistry
Unconjugated
Rabbit
Human
Cytosolic sialidase, EC 3.2.1.18, N-acetyl-alpha-neuraminidase 2, neuraminidase 2, SIAL2MGC129579, sialidase 2 (cytosolic sialidase), sialidase-2
The immunogen is a synthetic peptide directed towards the following sequence SGAHAYRIPALLYLPGQQSLLAFAEQRASKKDEHAELIVLRRGDYDAPTH (NP_005374). Peptide sequence SGAHAYRIPALLYLPGQQSLLAFAEQRASKKDEHAELIVLRRGDYDAPTH
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit