Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEU4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523889
Description
NEU4 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
NEU4 | |
Polyclonal | |
Unconjugated | |
NEU4 | |
9330166I04; LP5125; N-acetyl-alpha-neuraminidase 4; Neu4; Neuraminidase 4; neuraminidase 4 (sialidase); sialidase 4; sialidase-4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
129807, 241159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8BZL1, Q8WWR8 | |
NEU4 | |
KLH conjugated synthetic peptide between 423-452 amino acids from the C-terminal region of human NEU4. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction