Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEU4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557936
Description
Immunogen sequence: PRDDSWSVGP RSPLQPPLLG PGVHEPPEEA AVDPRGGQVP GGPFSRLQPR GDGPRQPGPR PGVSGDVGSW TLALPMPFAA PPQSPTWLLY SHPV Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 40%.
The protein encoded by this gene belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
NEU4 | |
Polyclonal | |
Unconjugated | |
NEU4 | |
9330166I04; LP5125; N-acetyl-alpha-neuraminidase 4; Neu4; Neuraminidase 4; neuraminidase 4 (sialidase); sialidase 4; sialidase-4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
129807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWR8 | |
NEU4 | |
Recombinant protein corresponding to Human NEU4. Recombinant protein control fragment (Product #RP-96236). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction