Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuN Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 13E6, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen NeuN
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB37725UMI
View Documents
MilliporeSigma
ZRB37725UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB3774X2MI
View Documents
MilliporeSigma
ZRB3774X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

NeuN Monoclonal antibody specifically detects NeuN in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

NeuN
Unconjugated
Rabbit
RNA binding protein fox-1 homolog 3;Fox-1 homolog C;Fox-3;Hexaribonucleotide-binding protein 3;NeuN;Neuronal nuclei
Monoclonal
Protein A Purified
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
GST/His-tagged recombinant fragment corresponding to the first 97 amino acids from the N-terminal region of murine Neuronal nuclei antigen (NeuN).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit