Learn More
Neuregulin-1/NRG1 Mouse anti-Human, Alexa Fluor™ 350, Clone: 19R23, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222030AF350
Catalog No. NP222030A30
Add to cart
Description
Description
Neuregulin-1/NRG1 Monoclonal antibody specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Neuregulin-1/NRG1
19R23
Western Blot, ELISA
ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
NRG1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant NRG1-b1/HRG1-b1 EFG Domain. Mouse anti human NRG1 beta-1/HRG1 beta-1 AAA58639.
0.1 mL
Primary
3084