Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuregulin-1/NRG1 Mouse anti-Human, Clone: 19R23, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Neuregulin-1/NRG1 |
---|---|
Clone | 19R23 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, ELISA 1:1000-2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP222030
|
Novus Biologicals
NBP222030 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019292
|
Novus Biologicals
NBP2220300.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Neuregulin-1/NRG1 Monoclonal antibody specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Neuregulin-1/NRG1 | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
AAA58639 | |
3084 | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant NRG1-b1/HRG1-b1 EFG Domain. Mouse anti human NRG1 beta-1/HRG1 beta-1 AAA58639. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
19R23 | |
Western Blot 1:500-1:1000, ELISA 1:1000-2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF | |
NRG1 | |
IgG1 | |
Protein G purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title