Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuregulin-1/NRG1 Mouse anti-Human, Mouse, Rat, Clone: 7D5, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85
Specifications
Antigen | Neuregulin-1/NRG1 |
---|---|
Clone | 7D5 |
Dilution | Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019407
|
Novus Biologicals
NBP2264540.05ML |
0.05 mL |
Each for $176.85
|
|
NBP226454
|
Novus Biologicals
NBP226454 |
0.25 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Neuregulin-1/NRG1 Monoclonal specifically detects Neuregulin-1/NRG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Neuregulin-1/NRG1 | |
Immunohistochemistry-Paraffin 1:50-1:100 | |
Monoclonal | |
Mouse | |
Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
PBS with BSA with 0.05% Sodium Azide | |
3084 | |
BALB/C mice were injected with a recombinant extracellular domain of rat NDF protein. | |
Primary | |
Store at 4C. Do not freeze. |
7D5 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Rat, Human, Mouse | |
ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF | |
NRG1 | |
IgG2a | |
Affinity Purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title