Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuritin Goat anti-Human, Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF283SP
This item is not returnable.
View return policy
Description
Neuritin Polyclonal specifically detects Neuritin in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Neuritin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dJ380B8.2, neuritin, neuritin 1, NRN1, NRNMGC44811 | |
Goat | |
Affinity Purified | |
RUO | |
51299 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q9NPD7 | |
NRN1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Neuritin Ala28-Gly114 Accession # Q9NPD7 | |
25 μg | |
Primary | |
Detects human Neuritin in direct ELISAs and Western blots. | |
Human, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only