Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEURL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599662
Description
Antibody detects endogenous levels of total Neuralized-1.
NEURL is involved in the determination of cell fate in the neurogenic region of the embryo and plays a role in the determination of cell fate in the central nervous system. NEURL may act as a tumor suppressor whose inactivation could be associated with malignant progression of astrocytic tumors.
Specifications
NEURL1 | |
Polyclonal | |
Unconjugated | |
NEURL1 | |
bA416N2.1; E3 ubiquitin-protein ligase NEURL1; h neu; h neuralized 1; h-neu; h-neuralized 1; neu; neu-1; NEUR1; neuralized E3 ubiquitin protein ligase 1; neuralized homolog; neuralized homolog 1A; neuralized-like homolog; neuralized-like protein 1A; NEURL; Neurl1; NEURL1A; RING finger protein 67; RING-type E3 ubiquitin transferase NEURL1; RNF67 | |
Rabbit | |
Affinity chromatography | |
RUO | |
9148 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O76050 | |
NEURL1 | |
A synthesized peptide derived from human NEURL1(Accession O76050), corresponding to amino acid residues S218-A268. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction