Learn More
Aviva Systems Biology Corp NEURL2 Antibody - N-terminal region (ARP52524_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52524_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEURL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEURL2. Alternate names: C20orf163, MGC125934, MGC125935, OZZ, Ozz-E3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60761