Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEURL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen NEURL3
Concentration 0.41 mg/mL
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-1085
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
166481AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

E3 ubiquitin-protein ligase. Seems to utilize UBE2E1. In vitro, generates polyubiquitin chains via non-canonical lysine residues suggesting that it is not involved in tagging substrates for proteasomal degradation.
Specifications

Specifications

NEURL3
Immunoprecipitation
Unconjugated
Rabbit
Human, Mouse
IgG
-20°C
0.41 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit