Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
NEURL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166481AP
Catalog No. 50-173-1085
Description
Description
E3 ubiquitin-protein ligase. Seems to utilize UBE2E1. In vitro, generates polyubiquitin chains via non-canonical lysine residues suggesting that it is not involved in tagging substrates for proteasomal degradation.
Specifications
Specifications
NEURL3
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CJC5, Q96EH8
LINCR, NEURL3
Rabbit
IgG
150 μL
-20°C
Primary
214854, 93082
Immunoprecipitation
Unconjugated
NEURL3
Liquid
NEURL3
NEURL3 Fusion Protein Ag10035
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse