Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurochondrin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259115

Catalog No. NBP259115


Add to cart

Description

Description

Neurochondrin Polyclonal antibody specifically detects Neurochondrin in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

Neurochondrin
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
KIAA0607NCDN-1, NCDN-2, neurochondrin
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Immunoprecipitation
Immunoprecipitation
Affinity Purified
NCDN
Maps to a region between 679 and 729
Immunogen affinity purified
RUO
Primary
23154
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only