Learn More
NeuroD1 Mouse anti-Human, Mouse, Alexa Fluor 488, Clone: 6E9G12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237415AF488
Catalog No. NBP237415Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
NeuroD1
6E9G12
Western Blot, ELISA
Purified
NEUROD1
Purified recombinant fragment of human Neurod (AA: 26-91) expressed in E. Coli.
Protein A or G purified
Neuroscience, Stem Cells
Primary
4760
ELISA, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
basic helix-loop-helix transcription factor, BETA2, BHF-1, BHLHA3, bHLHa3beta-cell E-box transactivator 2, Class A basic helix-loop-helix protein 3, MODY6, NeuroD, NeuroD1, neurogenic differentiation 1, neurogenic differentiation factor 1, neurogenic helix-loop-helix protein NEUROD, NIDDM
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse