Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuroD1 Mouse anti-Human, Mouse, Biotin, Clone: 6E9G12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237415B

Catalog No. NBP237415B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

NeuroD1
6E9G12
Western Blot, ELISA
Purified
NEUROD1
Purified recombinant fragment of human Neurod (AA: 26-91) expressed in E. Coli.
Protein A or G purified
Neuroscience, Stem Cells
Primary
4760
ELISA, Western Blot
Biotin
PBS with 0.05% Sodium Azide
basic helix-loop-helix transcription factor, BETA2, BHF-1, BHLHA3, bHLHa3beta-cell E-box transactivator 2, Class A basic helix-loop-helix protein 3, MODY6, NeuroD, NeuroD1, neurogenic differentiation 1, neurogenic differentiation factor 1, neurogenic helix-loop-helix protein NEUROD, NIDDM
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit