Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEUROD4 293T Cell Overexpression Lysate (Denatured), tested with Anti-NEUROD4 antibody (H00058158-B02), Abnova
Supplier: Abnova Corporation H00058158T01
Description
Western BlottingSpecifications
293T | |
NEUROD4 293T Cell Transient Overexpression Lysate(Denatured) | |
28.16 | |
pCMV-NEUROD4 full-length | |
100 μL | |
Human |
Western Blot | |
BC040961 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-NEUROD4 antibody (H00058158-B02) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction