Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurodye RH-160, pure, Acros Organics
Click to view available options
Quantity:
25mg
Specifications
Formula Weight | 470.66 |
Grade | Extra Pure |
Quantity | 25mg |
Chemical Name or Material | Neurodye RH-160 |
Safety and Handling
LIGHT SENSITIVE