Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurodye WW781, Acros Organics

Click to view available options
Quantity:
25mg
336185-19-4
C39H62N6O7S
759.02
MFCD12912451
CZJOQEYKRZTNMC-UHFFFAOYSA-N
(1, 3-Dibutylbarbituric acid-(5))-(3-methyl-1-p-sulfophenyl-5-pyrazolon(4))pentamethineoxonol triethylamine salt,WW78
131877954
4-[4-[5-(1,3-dibutyl-2,4,6-trioxo-1,3-diazinan-5-ylidene)penta-1,3-dienyl]-3-methyl-5-oxo-4H-pyrazol-1-yl]benzenesulfonic acid;N,N-diethylethanamine
CCCCN1C(=O)C(=CC=CC=CC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)O)C)C(=O)N(C1=O)CCCC.CCN(CC)CC.CCN(CC)CC
336185-19-4
C39H62N6O7S
759.02
MFCD12912451
CZJOQEYKRZTNMC-UHFFFAOYSA-N
(1, 3-Dibutylbarbituric acid-(5))-(3-methyl-1-p-sulfophenyl-5-pyrazolon(4))pentamethineoxonol triethylamine salt,WW78
131877954
4-[4-[5-(1,3-dibutyl-2,4,6-trioxo-1,3-diazinan-5-ylidene)penta-1,3-dienyl]-3-methyl-5-oxo-4H-pyrazol-1-yl]benzenesulfonic acid;N,N-diethylethanamine
CCCCN1C(=O)C(=CC=CC=CC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)O)C)C(=O)N(C1=O)CCCC.CCN(CC)CC.CCN(CC)CC
Formula Weight 759.01
Quantity 25mg
Chemical Name or Material Neurodye WW781