Learn More
Neurofascin Mouse anti-Human, Alexa Fluor 350, Clone: 1018843, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8208U100UG
Catalog No. FAB8208U100
Add to cart
This item is not returnable. View return policy
Description
Description
Neurofascin Monoclonal specifically detects Neurofascin in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Neurofascin
1018843
Flow Cytometry 0.25-1 ug/10^6 cells
NP_055905
DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NFASC, NRCAML
Human embryonic kidney cell, HEK293-derived human Neurofascin, Ile25-Trp1039, Accession # NP_055905
Protein A or G purified from cell culture supernatant
RUO
Primary
23114
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human