Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament Mouse anti-Human, Clone: 2F11, Abnova™

Mouse monoclonal antibody raised against human Neurofilament.

Manufacturer:  Abnova Corporation MAB12982

Catalog No. 89-934-686

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Neurofilament
2F11
Mouse monoclonal antibody raised against human Neurofilament.
In tissue culture supernatant (1% FCS, 10mM sodium azide).
Mouse
IgG1
RUO
Primary
Human
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Frozen sections) (1:160) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:20) The optimal working dilution should be determined by the end user.
Liquid
Human Neurofilament.
250 uL
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit