Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogenin 1 Mouse anti-Human, Clone: OTI15A7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524899

Catalog No. PIMA524899


Add to cart

Description

Description

Basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
Specifications

Specifications

Neurogenin 1
Monoclonal
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92886
Neurogenin 1, AKA, Math4C, NEUROD3, ngn1, bHLHa6
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4762
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
OTI15A7
Unconjugated
NEUROG1
Liquid
NEUROG1
Full-length protein expressed in 293T cell transfected with human NEUROG1 expression vector
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit