Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-1 Mouse anti-Human, Mouse, Rat, HRP, Clone: OTI3F9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272948H
Description
Neurogenin-1 Monoclonal specifically detects Neurogenin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Neurogenin-1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NEUROG1 | |
Full length recombinant protein of human Neurogenin-1 (NP_006152) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3F9 | |
Western Blot, Immunohistochemistry | |
AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1 | |
Mouse | |
Affinity Purified | |
Neuroscience | |
4762.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title