Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroligin 1 Mouse anti-Human, Mouse, Rat, Clone: S97A-31, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527635

Catalog No. PIMA527635


Add to cart

Description

Description

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
Specifications

Specifications

Neuroligin 1
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q62765, Q8N2Q7, Q99K10
NLG 1, KIAA1070, MGC45115, Neuroligin-1, NL1, NLG1, Nlgn1
Mouse
IgG1
100 μg
-20°C
Primary
116647, 192167, 22871
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S97A-31
Unconjugated
NLGN1
Liquid
Nlgn1
Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit