Learn More
Neuroligin 1/NLGN1 Mouse anti-Human, Mouse, Rat, Biotin, Clone: S97A-31, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242192B
Catalog No. NBP242192B
Add to cart
Description
Description
Neuroligin 1/NLGN1 Monoclonal antibody specifically detects Neuroligin 1/NLGN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Neuroligin 1/NLGN1
S97A-31
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
NLGN1
Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3.
Protein G purified
Cognition and Behavior
Primary
22871
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Greater then 40% identity with Neuroligin-2 and -3.Detects approx 120kDa. Does not cross-react with other Neuroligins.