Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroligin 1/NLGN1 Antibody (S97A-31), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242192FR
Description
Neuroligin 1/NLGN1 Monoclonal specifically detects Neuroligin 1/NLGN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Neuroligin 1/NLGN1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NLGN1 | |
Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3. | |
0.1 mL | |
Primary | |
Greater then 40% identity with Neuroligin-2 and -3.Detects approx 120kDa. Does not cross-react with other Neuroligins. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S97A-31 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1 | |
Mouse | |
Protein G purified | |
Cognition and Behavior | |
22871 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction