Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroligin 1 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$221.00 - $430.00
Specifications
Antigen | Neuroligin 1 |
---|---|
Clone | 001 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543128
![]() |
Thermo Scientific
MA543128 |
100 μL |
Each for $430.00
|
|
PIMA543433
![]() |
Thermo Scientific
MA543433 |
50 μL |
Each for $221.00
|
|
Description
Specificity: Human Neuroligin 1/NLGN1
Neuroligin 1 Recombinant Monoclonal antibody specifically detects Neuroligin 1 in Human samples. It is validated for ELISASpecifications
Neuroligin 1 | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
6330415N05Rik; BB179718; Kiaa1070; MGC45115; mKIAA1070; neuroligin 1; neuroligin I; Neuroligin-1; NL1; NLG 1; Nlg1; NLGN1 | |
Nlgn1 | |
IgG | |
Protein A | |
Antibody |
001 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
Q8N2Q7 | |
22871 | |
Recombinant Human Neuroligin 1/NLGN1 protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Nlgn1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title