Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat Neuroligin 2/NLGN2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF5645SP
This item is not returnable.
View return policy
Description
Neuroligin 2/NLGN2 Polyclonal specifically detects Neuroligin 2/NLGN2 in Human, Rat samples. It is validated for Western Blot.Specifications
Neuroligin 2/NLGN2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2, NLGN2 | |
Goat | |
Affinity Purified | |
RUO | |
57555 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1-3 ug/mL | |
EAW90197 | |
NLGN2 | |
Mouse myeloma cell line NS0-derived recombinant human Neuroligin 2/NLGN2 Gln15-Ser660 Accession # EAW90197 | |
25 μg | |
Primary | |
Detects human and rat Neuroligin 2/NLGN2 in direct ELISAs and Western blots. | |
Human, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction