Learn More
Neuroligin 4X/NLGN4X Mouse anti-Mouse, DyLight 405, Clone: S98-7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242198V
Catalog No. NBP242198V
Add to cart
Description
Description
Neuroligin 4X/NLGN4X Monoclonal antibody specifically detects Neuroligin 4X/NLGN4X in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Neuroligin 4X/NLGN4X
S98-7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
NLGN4X
Fusion protein amino acids 782-945 (intracellular C-terminus) of mouse Neuroligin 4. ~45% identity with Neuroligin-1, -2 and -3.
Protein G purified
Store at 4C in the dark.
Monoclonal
Approx 45% identity with Neuroligin-1, -2 and -3.Detects approx 25-130kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
ASPGX2, HLNX, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NLGN, NLGN4neuroligin 4, X-linked
Mouse
IgG1
0.1 ml
Primary
57502
Mouse