Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroligin 4Y/NLGN4Y Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen Neuroligin 4Y/NLGN4Y
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB028178
View Documents
Novus Biologicals
NBP293145100UL
100 μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NB028179
View Documents
Novus Biologicals
NBP29314520UL
20 μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Neuroligin 4Y/NLGN4Y Polyclonal antibody specifically detects Neuroligin 4Y/NLGN4Y in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Neuroligin 4Y/NLGN4Y
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
22829
IgG
Affinity purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
neuroligin 4, Y-linked, Neuroligin Y, neuroligin-4, Y-linked, Y linked
Recombinant fusion protein containing a sequence corresponding to amino acids 122-256 of human NLGN4Y (NP_001157710.1).
Primary
Store at -20°:C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit