Learn More
Tocris Bioscience™ Neuromedin N
Neurotensin-like peptide
Manufacturer:  Tocris Bioscience™ 1907/1
Catalog No. 19-071-R
Add to cart
This item is not returnable. View return policy
Description
Description
Endogenous neurotensin-like neuropeptide, originally isolated from porcine spinal cord. Binds to neurotensin receptors (IC50 = 16.7 nM for inhibition of [Trp11]-NT binding to rat brain receptors). Regulates guinea pig intestinal smooth muscle contraction and produces hypotension in rats. Also induces hypothermia following central administration in rats in vivo.
Specifications
Specifications
Neuromedin N
1mg
Neuromedin N (rat, mouse, porcine, canine)
745.96
Lys-Ile-Pro-Tyr-Ile-Leu