Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Neuromedin U-8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575470
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Neuromedin U (NmU) is a vertebrate peptide which stimulates uterine smooth muscle contraction and causes selective vasoconstriction. Like most other active peptides, it is proteolytically processed from a larger precursor protein. The mature peptides are 8 (NmU-8) to 25 (NmU-25) residues long and C-terminally amidated. The sequence of the C-terminal extremity of NmU is extremely well conserved across species.
Specifications
Neuromedin U-8 | |
Polyclonal | |
Unconjugated | |
NMU | |
Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; neuromedin U; neuromedin-U; Neuromedin-U-23; Neuromedin-U-25; Nmu; NmU 25; NmU-23; NmU-25; NURP33; NURP36; prepro-NMU | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10874, 56183, 63887 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P12760, P48645, Q9QXK8 | |
NMU | |
Synthetic peptide corresponding to amino acids N-terminus of Human Neuromedin U-8. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction