Learn More
neuron navigator 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 208071AP
Catalog No. 50-173-3099
Add to cart
Description
Description
NAV1 belongs to the neuron navigator family and is expressed predomitly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. The exact function of this gene is not known, but it is thought to play a role in in neuronal development and regeneration. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
neuron navigator 1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CH77, Q8NEY1
KIAA1151, KIAA1213, NAV1, neuron navigator 1, POMFIL3, steerin 1, STEERIN1, Unc 53 homolog 1, unc53H1
Rabbit
IgG
150 μL
-20°C
Primary
215690, 685707, 89796
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NAV1
Liquid
NAV1
neuron navigator 1 Fusion Protein Ag14747
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat