Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuronal Pentraxin 1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Neuronal Pentraxin 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179764
|
Novus Biologicals
NBP179764 |
100 μL |
Each of 1 for $436.00
|
|
Description
Neuronal Pentraxin 1 Polyclonal specifically detects Neuronal Pentraxin 1 in Mouse samples. It is validated for Western Blot.Specifications
Neuronal Pentraxin 1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DKFZp686J2446, MGC105123, neuronal pentraxin INP1NP-I, neuronal pentraxin-1 | |
NPTX1 | |
IgG | |
Affinity Purified | |
47 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_032756 | |
4884 | |
The immunogen for this antibody is Nptx1. Peptide sequence EGKGGPKNDTEERAKIESALTSLHQRISELEKGQKDNRPGDKFQLTFPLR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title