Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuronal Pentraxin 2, Mouse anti-Human, Clone: 569706, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems MAB7816100

 View more versions of this product

Catalog No. MAB7816100


Add to cart
This item is not returnable. View return policy

Description

Description

Neuronal Pentraxin 2 Monoclonal antibody specifically detects Neuronal Pentraxin 2 in Human Samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

Neuronal Pentraxin 2
569706
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL
P47972
Apexin, NARP, neuronal activity-regulated pentaxin, neuronal pentraxin IIapexin, neuronal pentraxin-2, NP2, NP-II, NPTX2
Mouse myeloma cell line, NS0-derived human Neuronal Pentraxin 2, Gly16-Leu431, Accession # P47972
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4885
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Purified
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Monoclonal
Detects human Neuronal Pentraxin 2 in direct ELISAs.
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit