Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropeptide FF R1/NPFFR1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300168SS
Description
Neuropeptide FF R1/NPFFR1 Polyclonal specifically detects Neuropeptide FF R1/NPFFR1 in Human samples. It is validated for Western Blot.Specifications
Neuropeptide FF R1/NPFFR1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NPFFR1 | |
N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein. | |
0.025 mg | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
64106 | |
Human | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 1:500 -1:1000 | |
FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Reacts with the N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction