Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide FF R1/NPFFR1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen Neuropeptide FF R1/NPFFR1
Concentration 1 mg/mL
Dilution Western Blot 1:500 -1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018147
SDP
View Documents
Novus Biologicals
NB300168SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB300168
SDP
View Documents
Novus Biologicals
NB300168
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

Neuropeptide FF R1/NPFFR1 Polyclonal specifically detects Neuropeptide FF R1/NPFFR1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Neuropeptide FF R1/NPFFR1
Western Blot 1:500 -1:1000
Polyclonal
Rabbit
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS with 0.02% Sodium Azide
64106
N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022
NPFFR1
IgG
Affinity Purified
Reacts with the N terminal sequence MEGEPSQPPNSSWPLS and the C terminal sequence CSHLPLTIPAWDI of the human NPFF1 protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.