Learn More
Tocris Bioscience™ Neuropeptide S (human)
Potent endogenous neuropeptide S receptor agonist
Supplier:  Tocris Bioscience™ 2587/1
Catalog No. 25871
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
Potent endogenous neuropeptide S receptor agonist (EC50 = 9.4 nM). Increases locomotor activity and wakefulness in mice. Also reduces anxiety-like behavior in mice.
Specifications
Specifications
Human
Store at -20°C
Observed M.W.: 2187.5
C93H155N31O28S
412938-67-1
Additional Peptide Receptor Products
1 mg