Learn More
Alfa Aesar™ Neuropeptide W-23, Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66112EXD
 View more versions of this product
Catalog No. AAJ66112EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2584.04
Human
Solid
RUO
Neuropeptide W-23, human
HPLC
0.5mg
Trp-Tyr-Lys-His-Val-Ala-Ser-Pro-Arg-Tyr-His-Thr-Val-Gly-Arg-Ala-Ala-Gly-Leu-Leu-Met-Gly-Leu
-30° to -10°C