Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropeptide Y Goat, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 13 publications
$206.00 - $436.00
Specifications
Antigen | Neuropeptide Y |
---|---|
Dilution | Western Blot 1: 300-1: 3000, Immunohistochemistry 1:300-1:3000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1: 300-1: 3000, Immunohistochemistry-Frozen 1: 300-1: 3000, Immunohistochemistry Free-Floating |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry - Free Floating |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP146535SS
|
Novus Biologicals
NBP146535SS |
0.05 mL |
Each for $206.00
|
|
NBP146535
|
Novus Biologicals
NBP146535 |
0.15 mL |
Each for $436.00
|
|
Description
Neuropeptide Y Polyclonal specifically detects Neuropeptide Y in Human, Mouse, Rat, Rabbit, Xenopus, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Specifications
Neuropeptide Y | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry - Free Floating | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Rabbit, Xenopus, Zebrafish | |
P57774 | |
4852 | |
A synthetic peptide from aa region 20-70 of mouse NPY (Neuropeptide Y) conjugated to an immunogenic carrier protein was used as the immunogen. | |
Primary | |
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with 0.15 ml sterilized water. | |
Specific for NPY. |
Western Blot 1: 300-1: 3000, Immunohistochemistry 1:300-1:3000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1: 300-1: 3000, Immunohistochemistry-Frozen 1: 300-1: 3000, Immunohistochemistry Free-Floating | |
Polyclonal | |
Goat | |
GPCR, Neuroscience, Neurotransmission | |
Whole antisera with No Preservative | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase | |
NPY | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title