Learn More
Enzo Life SciencesSupplier Diversity Partner Neuropeptide Y (h) monoclonal antibody (8) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner BPDABS0280802
Clone: 8. Host: Mouse. Isotype: IgG1κ. Immunogen: Synthetic human/rat neuropeptide Y. UniProt: P01303 (human). Species Reactivity: Human, Mouse, Rat. Applications: ELISA, IF, IHC. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-658-885