Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuropeptide Y Mouse, HRP, Clone: 8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105166H
Description
Neuropeptide Y Monoclonal specifically detects Neuropeptide Y in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Neuropeptide Y | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NPY | |
Synthetic human/rat NPY coupled to carrier with glutaraldehyde | |
0.1 mL | |
GPCR, Neuroscience, Neurotransmission | |
4852 | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
8 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Binds full length human/rat NPY. Cross-reactivity with NPY (1-24), NPY (18-36), PYY or PP was <0.2%. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title