Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuropeptideY Mouse anti-Human, Janelia Fluor 549, Clone: 8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP105166JF549

Catalog No. NBP15166549


Add to cart

Description

Description

NeuropeptideY Monoclonal antibody specifically detects NeuropeptideY in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

NeuropeptideY
8
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
NPY
Synthetic human/rat NPY coupled to carrier with glutaraldehyde
Protein A or G purified
GPCR, Neuroscience, Neurotransmission
Primary
4852
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Binds full length human/rat NPY. Cross-reactivity with NPY (1-24), NPY (18-36), PYY or PP was <0.2%.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit