Learn More
Neuropilin-1 (CD304) Mouse anti-Human, BB515, Clone: U21-1283  , BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566037
Catalog No. BDB566037
Add to cart
Description
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
Neuropilin-1 (CD304)
U21-1283  
Vol. Per test: 5μL
Purified
Murine
Mouse IgG2b, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R
Recombinant Human Neuropilin-1
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.