Learn More
Neuropilin-2 Mouse anti-Rat, Alexa Fluor 350, Clone: 96009R, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB567RU100UG
Catalog No. FAB567RU100
Add to cart
This item is not returnable. View return policy
Description
Description
Neuropilin-2 Monoclonal specifically detects Neuropilin-2 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
Neuropilin-2
96009R
Flow Cytometry 0.25-1 ug/106 cells
O35276
MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17)
Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2, Gln23-Asp857 (Val809-Asp825 del), Accession # O35276
Protein A or G purified from cell culture supernatant
RUO
Primary
8828
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG2b
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Rat