Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Neuroplastin Monoclonal Antibody (OTI2D4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525884
Description
Neuroplastin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
Specifications
Neuroplastin | |
Monoclonal | |
0.69 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97300, Q9Y639 | |
NPTN | |
Full length human recombinant protein of NPTN produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Canine, Monkey | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2D4 | |
Unconjugated | |
NPTN | |
AW554172; glycoprotein 55; glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; Sdfr1; Sdr1; SDR-1; stromal cell derived factor receptor 1; stromal cell-derived receptor 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
20320, 27020, 487637 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction