Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroplastin Mouse anti-Human, Rat, Canine, Monkey, DyLight™ 488, Clone: OTI3C4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272966G
Description
Neuroplastin Monoclonal specifically detects Neuroplastin in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Neuroplastin | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NPTN | |
Full length recombinant protein of human Neuroplastin (NP_059429) produced in HEK293T cells. | |
0.1 mL | |
27020.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3C4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
GP55Stromal cell-derived receptor 1, GP65SDR-1, MGC102805, neuroplastin, np55, np65, SDFR1DKFZp686L2477, SDR1stromal cell derived factor receptor 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat, Canine, Monkey | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title